4-(Trifluoromethoxy)phenyl isothiocyanate
Leverantör: Thermo Scientific
Synonyms: 4-(Trifluoromethoxy)phenylisothiocyanate
TL00411DA TL00411EA
MAYBTL00411DAEA 0 SEK
MAYBTL00411DA MAYBTL00411EA
4-(Trifluoromethoxy)phenyl isothiocyanate
4-(Trifluoromethoxy)phenyl isothiocyanate
Formel: C₈H₄F₃NOS
MW: 219,19 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00042412
CAS nummer: 64285-95-6