2-(Trifluoromethoxy)phenyl isothiocyanate

Leverantör: Thermo Scientific

SB01674EA
MAYBSB01674EAEA 0 SEK
MAYBSB01674EA
2-(Trifluoromethoxy)phenyl isothiocyanate
2-(Trifluoromethoxy)phenyl isothiocyanate
Formel: C₈H₄F₃NOS
MW: 219,19 g/mol
MDL Number: MFCD00220860
CAS nummer: 175205-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR