2,4-Difluorophenyl isocyanate ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: 2,4-Difluorophenylisocyanate
A12415.06 A12415.14 A12415.22
A12415.06EA 1125 SEK
A12415.06 A12415.14 A12415.22
2,4-Difluorophenyl isocyanate ≥98%
2,4-Difluorophenyl isocyanate
Purity: 98+% CAS No.: 59025-55-7 Molecular Formula: C7H3F2NO Formula Weight: 155.11 Flash Point: 50°(122°F) Refractive Index: 1.4920 Boiling Point: 64-66°/10mm Beilstein Reference: 2831577 UN No.: 3080 MDL No.: MFCD00001997
Formel: C₇H₃F₂NO
MW: 155,1 g/mol
Kokpunkt: 64…66 °C (10 mmHg)
Densitet: 1,310
Flampunkt: 50 °C (122 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00001997
CAS nummer: 59025-55-7
UN: 3080
ADR: 6.1,II