1-Iodo-4-(trifluoromethoxy)benzene ≥98%
Leverantör: Thermo Fisher Scientific
|
Varning
|
Synonyms:
4-(Trifluoromethoxy)iodobenzene
B23628.03
B23628.06
B23628.14
B23628.03EA
295
SEK
B23628.03
B23628.06
B23628.14
1-Iodo-4-(trifluoromethoxy)benzene ≥98%
1-Iodo-4-(trifluoromethoxy)benzene
Purity: 98%
CAS No.: 103962-05-6
Molecular Formula: C7H4F3IO
Formula Weight: 288.01
Flash Point: 77°(170°F)
Refractive Index: 1.5033
Boiling Point: 72-74°/15mm
Beilstein Reference: 6193794
MDL No.: MFCD00042411
Formel:
C₇H₄F₃IO MW: 288.01 g/mol Kokpunkt: 72…74 °C (15 mmHg) Densitet: 1.909 Flampunkt: 77 °C (170 °F) Storage Temperature: Omgivande |
MDL Number:
MFCD00042411 CAS nummer: 103962-05-6 |