Ethyl-4-fluorobenzoate ≥99%
Leverantör: Thermo Scientific
Synonyms: 4-Fluorobenzoic acid ethyl ester, Ethyl 4-fluorobenzoate
B21484.06 B21484.14 B21484.22
B21484.06EA 665 SEK
B21484.06 B21484.14 B21484.22
Ethyl-4-fluorobenzoate ≥99%
Ethyl-4-fluorobenzoate
Purity: 99% CAS No.: 451-46-7 Molecular Formula: C9H9FO2 Formula Weight: 168.17 Flash Point: 81°(177°F) Refractive Index: 1.4870 Odour: Recognizable Melting Point: 25-27° Boiling Point: 209-210° Beilstein Reference: 1866794 Merck Reference: 14,4171 MDL No.: MFCD00000351
Formel: C₉H₉FO₂
MW: 168,17 g/mol
Kokpunkt: 209…210 °C
Smältpunkt: 25…27 °C
Densitet: 1,146
Flampunkt: 81 °C (177 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000351
CAS nummer: 451-46-7
EINECS: 207-194-8
Merck Index: 14,4171