Ethyl-3-fluorobenzoate ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: Ethyl 3-fluorobenzoate, 3-Fluorobenzoic acid ethyl ester
L07984.09 L07984.18
L07984.09EA 1000 SEK
L07984.09 L07984.18
Ethyl-3-fluorobenzoate ≥98%
Ethyl-3-fluorobenzoate
Purity: 98+% CAS No.: 451-02-5 Molecular Formula: C9H9FO2 Formula Weight: 168.17 Flash Point: 82°(179°F) Refractive Index: 1.4850 Odour: Recognizable Boiling Point: 208° Beilstein Reference: 2045876 MDL No.: MFCD00000336
Formel: C₉H₉FO₂
MW: 168,17 g/mol
Kokpunkt: 208 °C
Densitet: 1,136
Flampunkt: 82 °C (179 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000336
CAS nummer: 451-02-5
EINECS: 207-191-1