3,5-Difluorobenzoyl chloride ≥98%
Leverantör: Thermo Scientific
Fara
B21400.06 B21400.14
B21400.06EA 1450 SEK
B21400.06 B21400.14
3,5-Difluorobenzoyl chloride ≥98%
3,5-Difluorobenzoyl chloride
Purity: 98+% CAS No.: 129714-97-2 Molecular Formula: C7H3ClF2O Formula Weight: 176.55 Flash Point: 69°(156°F) Refractive Index: 1.5030 Boiling Point: 173-175° Beilstein Reference: 4307685 UN No.: 3265 MDL No.: MFCD00010309
Formel: C₇H₃ClF₂O
MW: 176,55 g/mol
Kokpunkt: 173…175 °C
Densitet: 1,41
Flampunkt: 69 °C (156 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00010309
CAS nummer: 129714-97-2
UN: 3265
ADR: 8,II