3,4-Difluorobenzoyl chloride ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: 3,4-difluorobenzoylchloride
L08188.06 L08188.14
L08188.06EA 1375 SEK
L08188.06 L08188.14
3,4-Difluorobenzoyl chloride ≥98%
3,4-Difluorobenzoyl chloride
Purity: 98% CAS No.: 76903-88-3 Molecular Formula: C7H3ClF2O Formula Weight: 176.55 Flash Point: 78°(172°F) Refractive Index: 1.5110 Boiling Point: 185-186° Beilstein Reference: 4177619 UN No.: 3265 MDL No.: MFCD00009930
Formel: C₇H₃ClF₂O
MW: 176,55 g/mol
Kokpunkt: 185…186 °C
Densitet: 1,402
Flampunkt: 78 °C (172 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00009930
CAS nummer: 76903-88-3
EINECS: 278-570-7
UN: 3265
ADR: 8,II