2-Fluorobenzoyl chloride 97%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: o-Fluorobenzoyl chloride
153090250 153091000
ACRO153090250EA 665 SEK
ACRO153090250 ACRO153091000
2-Fluorobenzoyl chloride 97%
2-Fluorobenzoyl chloride
Formel: C₇H₄ClFO
Kokpunkt: 90…92 °C (15 mmHg)
Smältpunkt: 4 °C
Densitet: 1,32 g/cm³
Flampunkt: 82 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000656
CAS nummer: 393-52-2
EINECS: 206-887-2
UN: 1760
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Clear colorless to slightly colored Liquid
Infrared spectrum Conforms
Refractive index 1.5355 to 1.5375 (20°C, 589 nm)
GC ≥96.0 %