2,5-Difluorobenzoyl chloride ≥98%
Leverantör: Thermo Scientific
Fara
A10651.06 A10651.14
A10651.06EA 1125 SEK
A10651.06 A10651.14
2,5-Difluorobenzoyl chloride ≥98%
2,5-Difluorobenzoyl chloride
Purity: 98% CAS No.: 35730-09-7 Molecular Formula: C7H3ClF2O Formula Weight: 176.55 Flash Point: 93°(199°F) Refractive Index: 1.5150 Boiling Point: 81-83°/14mm Beilstein Reference: 2046666 UN No.: 3265 MDL No.: MFCD00009929
Formel: C₇H₃ClF₂O
MW: 176,55 g/mol
Kokpunkt: 81…83 °C (14 mmHg)
Densitet: 1,425
Flampunkt: 93 °C (199 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00009929
CAS nummer: 35730-09-7
EINECS: 252-701-8
UN: 3265
ADR: 8,II