4-Fluorophthalonitrile ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: 4-Fluorobenzene-1,2-dicarbonitrile, 4-Fluoro-1,2-benzenedicarbonitrile, 4-Fluoro-1,2-dicyanobenzene
B22624.03 B22624.06
B22624.03EA 2625 SEK
B22624.03 B22624.06
4-Fluorophthalonitrile ≥98%
4-Fluorophthalonitrile
Purity: 98% CAS No.: 65610-14-2 Molecular Formula: C8H3FN2 Formula Weight: 146.12 Melting Point: 100-102° Beilstein Reference: 2443236 UN No.: 3439 MDL No.: MFCD00210430
Formel: C₈H₃FN₂
MW: 146,12 g/mol
Kokpunkt: 294 °C (1013 hPa)
Smältpunkt: 100…102 °C
Storage Temperature: Omgivande
MDL Number: MFCD00210430
CAS nummer: 65610-14-2
UN: 3439
ADR: 6.1,II