4-(Trifluoromethoxy)benzaldehyde 96%
Leverantör: Thermo Fisher Scientific
Varning
A10671.14 A10671.06
A10671.14EA 3575 SEK
A10671.14 A10671.06
4-(Trifluoromethoxy)benzaldehyde 96%
4-(Trifluoromethoxy)benzaldehyde
Purity: 96% CAS No.: 659-28-9 Molecular Formula: C8H5F3O2 Formula Weight: 190.12 Flash Point: 71°(159°F) Refractive Index: 1.4580 Boiling Point: 93°/27mm Beilstein Reference: 1949135 MDL No.: MFCD00041530
Formel: C₈H₅F₃O₂
MW: 190.12 g/mol
Kokpunkt: 93 °C (27 mmHg)
Densitet: 1.331
Flampunkt: 71 °C (159 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00041530
CAS nummer: 659-28-9
EINECS: 211-531-4