Kaliumväteftalat 99+%
Leverantör: Thermo Scientific
Synonyms: Potassium phthalate monobasic, Potassium biphthalate, Potassium Acid Phthalate, KHP, Phthalic Acid Monopotassium Salt, Phthalic acid potassium salt
417955000 417950025
ACRO417955000EA 1875 SEK
ACRO417955000 ACRO417950025
Kaliumväteftalat 99+%
Kaliumväteftalat
Formel: HOOCC₆H₄COOK
MW: 204,22 g/mol
Smältpunkt: 295…300 °C (dec.)
Densitet: 1,6362 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00013070
CAS nummer: 877-24-7
EINECS: 212-889-4
Merck Index: 13,07694
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4652479e822d1591ac7eb5e29f892b8c94588ad4&vid=40ddd2ae91fd20c68037480d22b8fbe61e8f44e8&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Titration with NaOH ≥99.0 %
GC ≥97.5 %