Tereftalsyra ≥98%
Leverantör: Thermo Fisher Scientific
Varning
A12527.0C A12527.30 A12527.36
A12527.0CEA 14400 SEK
A12527.0C A12527.30 A12527.36
Tereftalsyra ≥98%
Tereftalsyra
Purity: 98+% CAS No.: 100-21-0 Molecular Formula: C8H6O4 Formula Weight: 166.13 Flash Point: 260°(500°F) Odour: Odourless Beilstein Reference: 1909333 Merck Reference: 14,9162 MDL No.: MFCD00002558
Formel: C₈H₆O₄
Kokpunkt: 392 °C (1013 hPa)
Smältpunkt: 402 °C (subl.)
Densitet: 1,510
Flampunkt: 260 °C (500 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00002558
CAS nummer: 100-21-0
EINECS: 202-830-0
Merck Index: 14,9162