Isophthalic acid 99
Leverantör: Thermo Fisher Scientific
A14445.0B A14445.0I A14445.30
A14445.0BEA 845 SEK
A14445.0B A14445.0I A14445.30
Isophthalic acid 99
Isophthalic acid
Purity: 99% CAS No.: 121-91-5 Molecular Formula: C8H6O4 Formula Weight: 166.13 Melting Point: 344-348° subl. Beilstein Reference: 1909332 Merck Reference: 14,5197 MDL No.: MFCD00002516
Formel: C₈H₆O₄
MW: 166.13 g/mol
Kokpunkt: 119…121 °C (6 mmHg)
Smältpunkt: 344…348 °C (subl.)
Densitet: 1.540
Storage Temperature: Omgivande
MDL Number: MFCD00002516
CAS nummer: 121-91-5
EINECS: 204-506-4
Merck Index: 13,05216