2-Ftalsyra ≥99.5% ACS
Leverantör: Thermo Fisher Scientific
Varning
41770.22 41770.36
41770.22EA 1075 SEK
41770.22 41770.36
2-Ftalsyra ≥99.5% ACS
2-Ftalsyra
Purity: 99.5+% CAS No.: 88-99-3 Molecular Formula: C6H4(COOH)2 Formula Weight: 166.13 Flash Point: 168°(334°F) Physical Form: Crystalline powder Density: 1.593 Appearance: White Odour: Odourless Melting Point: 205° dec. Beilstein Reference: 608199 Merck Reference: 14,7371 MDL No.: MFCD00002467
Formel: C₈H₆O₄
Kokpunkt: 315…317 °C (1013 hPa)
Smältpunkt: 205 °C (dec.)
Densitet: 1,593
Flampunkt: 168 °C (334 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00002467
CAS nummer: 88-99-3
EINECS: 201-873-2
Merck Index: 14,7371