3-(Methylthio)benzoic acid ≥97%
Leverantör: Thermo Fisher Scientific
Varning
H27200.03 H27200.06 H27200.14
H27200.03EA 1075 SEK
H27200.03 H27200.06 H27200.14
3-(Methylthio)benzoic acid ≥97%
3-(Methylthio)benzoic acid
Purity: 97% CAS No.: 825-99-0 Molecular Formula: C8H8O2S Formula Weight: 168.21 Melting Point: 126-132° MDL No.: MFCD00156993
Formel: C₈H₈O₂S
Smältpunkt: 126…132 °C
Storage Temperature: Omgivande
MDL Number: MFCD00156993
CAS nummer: 825-99-0