3,5-Dinitrosalicylsyra, Sigma-Aldrich®

Leverantör: SIGMA ALDRICH MICROSCOPY
Varning

Synonyms: 2-Hydroxy-3,5-dinitrobenzoic acid, 3,5-Dinitro-2-hydroxybenzoic acid

D0550-10G
D0550-10GEA 269 SEK
D0550-10G
3,5-Dinitrosalicylsyra, Sigma-Aldrich®
3,5-Dinitrosalicylsyra
Formel: C₇H₄N₂O₇
MW: 228.12 g/mol
Smältpunkt: 170…174 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007104
CAS nummer: 609-99-4
EINECS: 210-204-3
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c39003c66998ed33039f29dcf62cdb2a9d2b9aa&vid=bed33ffccf9a3dfe330b6ae1492067f2c779ec9f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR