Dimethyl terephthalate 99%
Leverantör: Thermo Fisher Scientific
180580010 180580025 180580250 180585000 180580000
ACRO180580010EA 1075 SEK
ACRO180580010 ACRO180580025 ACRO180580250 ACRO180585000 180580000.
Dimethyl terephthalate 99%
Dimethyl terephthalate
Formel: C₁₀H₁₀O₄
Kokpunkt: 288 °C (1013 hPa)
Smältpunkt: 140…143 °C
Densitet: 1,2 g/cm³ (20 °C)
Flampunkt: 154 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008440
CAS nummer: 120-61-6
EINECS: 204-411-8
Merck Index: 12,09306
Order Now
Specifikation testresultat
Appearance White Flakes and/or pellets
Infrared spectrum Conforms
Melting point 140 to 143 °C
GC ≥98.5 %
Acid value ≤0.03 mg KOH/g