Natrium 4-Hydroxipropylbensoat, EMPROVE® ESSENTIAL Ph. Eur., BP, NF, SAFC®
Leverantör: MERCK PRODUCTION CHEMICALS
Fara
Synonyms: Propyl parahydroxybenzoate sodium salt, Sodium Propyl Parahydroxybenzoate, Propylparaben Sodium, Propylparaben sodium salt, 4-Hydroxybenzoic acid propyl ester sodium salt, Sodium 4-(propoxycarbonyl)phenolate, Sodium 4-propoxycarbonylphenoxide
1.07428.5000 1.07428.9029
1.07428.5000EA 14125 SEK
1.07428.5000 1.07428.9029
Natrium 4-Hydroxipropylbensoat, EMPROVE® ESSENTIAL Ph. Eur., BP, NF, SAFC®
Natrium 4-Hydroxipropylbensoat
Formel: C₁₀H₁₁NaO₃
MW: 202.19 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00016477
CAS nummer: 35285-69-9
EINECS: 252-488-1