Propyl-4-hydroxibensoat ≥99%
Leverantör: Thermo Scientific
Varning
Synonyms: Propyl 4-hydroxybenzoate, Propyl parahydroxybenzoate, Propylparaben, 4-Hydroxybenzoic acid propyl ester
A10180.30 A10180.0B A10180.0I
A10180.30EA 1100 SEK
A10180.30 A10180.0B A10180.0I
Propyl-4-hydroxibensoat ≥99%
Propyl-4-hydroxibensoat
Purity: 99+% CAS No.: 94-13-3 Molecular Formula: C10H12O3 Formula Weight: 180.20 Flash Point: 180°(356°F) Melting Point: 95-99° Beilstein Reference: 1103245 Merck Reference: 14,7866 MDL No.: MFCD00002354
Formel: C₁₀H₁₂O₃
MW: 180,2 g/mol
Kokpunkt: 292 °C (1013 hPa)
Smältpunkt: 95…99 °C
Densitet: 1,28 g/cm³ (20 °C)
Flampunkt: 180 °C (356 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00002354
CAS nummer: 94-13-3
EINECS: 202-307-7
Merck Index: 14,7866