Butyl-4-hydroxibensoat ≥99%
Leverantör: Thermo Scientific
Synonyms: Butyl 4-hydroxybenzoate, 4-Hydroxybenzoic acid butyl ester, Butyl parahydroxybenzoate, Butylparaben
A14043.22 A14043.30
A14043.22EA 855 SEK
A14043.22 A14043.30
Butyl-4-hydroxibensoat ≥99%
Butyl-4-hydroxibensoat
Purity: 99+% CAS No.: 94-26-8 Molecular Formula: C11H14O3 Formula Weight: 194.23 Melting Point: 67-71° Beilstein Reference: 1103741 Merck Reference: 14,1584 MDL No.: MFCD00016478
Formel: C₁₁H₁₄O₃
MW: 194,23 g/mol
Kokpunkt: 156…157 °C (3,5 mmHg)
Smältpunkt: 67…71 °C
Densitet: 1,23 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00016478
CAS nummer: 94-26-8
EINECS: 202-318-7
Merck Index: 14,1584