Methyl-N-methylanthranilate ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Methyl N-Methylanthranilate, Dimethyl anthranilate
B20233.18 B20233.30
B20233.18EA 1125 SEK
B20233.18 B20233.30
Methyl-N-methylanthranilate ≥98%
Methyl-N-methylanthranilate
Purity: 98% CAS No.: 85-91-6 Molecular Formula: C9H11NO2 Formula Weight: 165.19 Flash Point: 133°(271°F) Refractive Index: 1.5800 Melting Point: 17-19° Boiling Point: 256-258° Beilstein Reference: 607217 MDL No.: MFCD00017183
Formel: C₉H₁₁NO₂
Kokpunkt: 256…258 °C
Smältpunkt: 17…19 °C
Densitet: 1,130
Flampunkt: 133 °C (271 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00017183
CAS nummer: 85-91-6
EINECS: 201-642-6