Methyl-3-aminobenzoate ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: Methyl 3-aminobenzoate
A14766.06 A14766.14 A14766.22
A14766.06EA 1275 SEK
A14766.06 A14766.14 A14766.22
Methyl-3-aminobenzoate ≥98%
Methyl-3-aminobenzoate
Purity: 98+% CAS No.: 4518-10-9 Molecular Formula: C8H9NO2 Formula Weight: 151.17 Melting Point: 49-53° Beilstein Reference: 1101468
Formel: C₈H₉NO₂
MW: 151,16 g/mol
Kokpunkt: 120 °C (0,7 mmHg)
Smältpunkt: 49…53 °C
Densitet: 1,232
Storage Temperature: Omgivande
MDL Number: MFCD00017102
CAS nummer: 4518-10-9
EINECS: 224-842-5