Ethyl-4-(dimethylamino)benzoate 99%
Leverantör: Thermo Scientific
Synonyms: 4-(Dimethylamino)benzoic acid ethyl ester, Ethyl 4-dimethylaminobenzoate
A12888.14 A12888.22 A12888.36
A12888.14EA 725 SEK
A12888.14 A12888.22 A12888.36
Ethyl-4-(dimethylamino)benzoate 99%
Ethyl-4-(dimethylamino)benzoate
Purity: 99% CAS No.: 10287-53-3 Molecular Formula: C11H15NO2 Formula Weight: 193.25 Melting Point: 63-65° Boiling Point: 190-191°/14mm MDL No.: MFCD00009115
Formel: C₁₁H₁₅NO₂
MW: 193,25 g/mol
Kokpunkt: 190…191 °C (14 mmHg)
Smältpunkt: 63…65 °C
Densitet: 1,060
Storage Temperature: Omgivande
MDL Number: MFCD00009115
CAS nummer: 10287-53-3
EINECS: 233-634-3