3,5-Dihydroxybenzhydrazide ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 3,5-Dihydroxybenzoic hydrazide
L11471.06 L11471.14
L11471.06EA 1325 SEK
L11471.06 L11471.14
3,5-Dihydroxybenzhydrazide ≥98%
3,5-Dihydroxybenzhydrazide
Purity: 98% CAS No.: 7732-32-3 Molecular Formula: C7H8N2O3 Formula Weight: 168.15 Melting Point: 256-257° Beilstein Reference: 2642968
Formel: C₇H₈N₂O₃
MW: 168,15 g/mol
Smältpunkt: 256…257 °C
Storage Temperature: Omgivande
MDL Number: MFCD00017066
CAS nummer: 7732-32-3