4-Butoxybenzoyl chloride ≥98%
Leverantör: Thermo Fisher Scientific
Fara
A11658.06 A11658.14
A11658.06EA 1600 SEK
A11658.06 A11658.14
4-Butoxybenzoyl chloride ≥98%
4-Butoxybenzoyl chloride
Purity: 98% CAS No.: 33863-86-4 Molecular Formula: C11H13ClO2 Formula Weight: 212.68 Flash Point: 110°(230°F) Refractive Index: 1.5495 Boiling Point: 161-162°/9mm Beilstein Reference: 957203 UN No.: 3265 MDL No.: MFCD00000689
Formel: C₁₁H₁₃ClO₂
Kokpunkt: 161…162 °C (9 mmHg)
Densitet: 1,121
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000689
CAS nummer: 33863-86-4
EINECS: 251-703-6
UN: 3265
ADR: 8,II