3,5-Dinitrobenzoylklorid ≥98%
Leverantör: Thermo Scientific
Fara
A16729.22 A16729.14
A16729.22EA 3425 SEK
A16729.22 A16729.14
3,5-Dinitrobenzoylklorid ≥98%
3,5-Dinitrobenzoylklorid
Purity: 98+% CAS No.: 99-33-2 Molecular Formula: C7H3ClN2O5 Formula Weight: 230.57 Melting Point: 67-70° Boiling Point: 196°/12mm Merck Reference: 14,3277 UN No.: 3261 MDL No.: MFCD00007248
Formel: C₇H₃ClN₂O₅
MW: 230,56 g/mol
Kokpunkt: 196 °C (12 mmHg)
Smältpunkt: 67…70 °C
Densitet: 1,33 g/cm³ (20 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00007248
CAS nummer: 99-33-2
EINECS: 202-750-6
UN: 3261
ADR: 8,II
Merck Index: 14,3277