4-Nitrobensoylklorid ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: p-Nitrobenzoyl chloride
A12543.09 A12543.0B A12543.30
A12543.09EA 580 SEK
A12543.09 A12543.0B A12543.30
4-Nitrobensoylklorid ≥98%
4-Nitrobensoylklorid
Purity: 98% CAS No.: 122-04-3 Molecular Formula: C7H4ClNO3 Formula Weight: 185.57 Flash Point: 102°(215°F) Odour: Characteristic Boiling Point: 155°/15mm Beilstein Reference: 473192 Merck Reference: 14,6590 UN No.: 3261 MDL No.: MFCD00007345
Formel: C₇H₄ClNO₃
MW: 185,57 g/mol
Kokpunkt: 155 °C (15 mmHg)
Smältpunkt: 71…74 °C
Densitet: 1,53
Flampunkt: 102 °C (215 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00007345
CAS nummer: 122-04-3
EINECS: 204-517-4
UN: 3261
ADR: 8,II
Merck Index: 14,6590