Phthalonitrile ≥98%

Leverantör: Thermo Scientific
Fara

Synonyms: 1,2-Benzenedicarbonitrile, Benzene-1,2-dicarbonitrile, 1,2-Dicyanobenzene

A15071.0B A15071.22 A15071.30
A15071.0BEA 2425 SEK
A15071.0B A15071.22 A15071.30
Phthalonitrile ≥98%
Phthalonitrile

Purity: 98% CAS No.: 91-15-6 Molecular Formula: C8H4N2 Formula Weight: 128.13 Flash Point: 162°(323°F) Melting Point: 139-141° Beilstein Reference: 775028 UN No.: 3439 MDL No.: MFCD00001771

Formel: C₈H₄N₂
MW: 128,13 g/mol
Kokpunkt: 305 °C (1013 hPa)
Smältpunkt: 139…141 °C
Densitet: 1,1 g/cm³ (20 °C)
Flampunkt: 162 °C (323 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00001771
CAS nummer: 91-15-6
EINECS: 202-044-8
UN: 3439
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR