Phthalonitrile 98%
Leverantör: Thermo Fisher Scientific
Fara
174010010 174012500
ACRO174010010EA 1725 SEK
ACRO174010010 ACRO174012500
Phthalonitrile 98%
Phthalonitrile
Formel: C₈H₄N₂
Kokpunkt: 305 °C (1013 hPa)
Smältpunkt: 138…142 °C
Densitet: 1,24 g/cm³
Flampunkt: >130 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001771
CAS nummer: 91-15-6
EINECS: 202-044-8
UN: 3439
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance White to beige to grey-green Powder or crystals
Infrared spectrum Conforms
Melting point 138 to 142 °C
GC ≥97.5 %