4-Cyanobenzenesulphonyl chloride ≥97%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 4-Cyanobenzene-1-sulphonyl chloride
A14081.14 A14081.03 A14081.06
A14081.14EA 15150 SEK
A14081.14 A14081.03 A14081.06
4-Cyanobenzenesulphonyl chloride ≥97%
4-Cyanobenzenesulphonyl chloride
Purity: 97% CAS No.: 49584-26-1 Molecular Formula: C7H4ClNO2S Formula Weight: 201.63 Melting Point: 107-110° Beilstein Reference: 777939 UN No.: 3261 MDL No.: MFCD00052337
Formel: C₇H₄ClNO₂S
MW: 201.63 g/mol
Smältpunkt: 107…110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00052337
CAS nummer: 49584-26-1
UN: 3261
ADR: 8 II