2,4,6-Trimethoxybenzonitrile ≥98%
Leverantör: Thermo Fisher Scientific
Varning
B23540.06 B23540.14
B23540.06EA 885 SEK
B23540.06 B23540.14
2,4,6-Trimethoxybenzonitrile ≥98%
2,4,6-Trimethoxybenzonitrile
Purity: 98% CAS No.: 2571-54-2 Molecular Formula: C10H11NO3 Formula Weight: 193.20 Odour: Characteristic Melting Point: 140-143° MDL No.: MFCD00001787
Formel: C₁₀H₁₁NO₃
Kokpunkt: 347 °C (1013 hPa)
Smältpunkt: 140…143 °C
Densitet: 1,36 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00001787
CAS nummer: 2571-54-2
EINECS: 219-917-4