Methyl-4-cyanobenzoate ≥99%
Leverantör: Thermo Scientific
Fara
Synonyms: 4-Cyanobenzoic acid methyl ester
L08365.06 L08365.14 L08365.22
L08365.06EA 700 SEK
L08365.06 L08365.14 L08365.22
Methyl-4-cyanobenzoate ≥99%
Methyl-4-cyanobenzoate
Purity: 99% CAS No.: 1129-35-7 Molecular Formula: C9H7NO2 Formula Weight: 161.16 Melting Point: 65-69° Boiling Point: 142-144°/12mm MDL No.: MFCD00001823
Formel: C₉H₇NO₂
MW: 161,16 g/mol
Kokpunkt: 142…144 °C (12 mmHg)
Smältpunkt: 65…69 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001823
CAS nummer: 1129-35-7
EINECS: 214-443-4
UN: 3439
ADR: 6.1,III