4-Chlorobenzenesulphonyl chloride 97%
Leverantör: Thermo Scientific
Fara
Synonyms: 4-Chlorobenzene-1-sulphonyl chloride
158560050 158560250 158561000 158565000
ACRO158560050EA 470 SEK
ACRO158560050 ACRO158560250 ACRO158561000 ACRO158565000
4-Chlorobenzenesulphonyl chloride 97%
4-Chlorobenzenesulphonyl chloride
Formel: C₆H₄Cl₂O₂S
MW: 211,07 g/mol
Kokpunkt: 272 °C (1013 hPa)
Smältpunkt: 50…52 °C
Flampunkt: 107 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007439
CAS nummer: 98-60-2
EINECS: 202-685-3
UN: 3261
ADR: 8,II
Order Now
Specifikation testresultat
Appearance White to light yellow Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 50 to 55 °C
Titration RCOCl ≥96.0 %
Titrable acid ≤0.002 meq/g