3-Chlorobenzenesulphonyl chloride ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: 3-Chlorobenzene-1-sulphonyl chloride
A10283.06 A10283.03 A10283.14
A10283.06EA 2150 SEK
A10283.06 A10283.03 A10283.14
3-Chlorobenzenesulphonyl chloride ≥98%
3-Chlorobenzenesulphonyl chloride
Purity: 98% CAS No.: 2888-06-4 Molecular Formula: C6H4Cl2O2S Formula Weight: 211.07 Flash Point: 110°(230°F) Refractive Index: 1.5690 Melting Point: ca 21° Boiling Point: 102-104°/1mm Beilstein Reference: 2937139 UN No.: 3265 MDL No.: MFCD00051697
Formel: C₆H₄Cl₂O₂S
MW: 211,07 g/mol
Kokpunkt: 102…104 °C (1 mmHg)
Smältpunkt: ∼ 21 °C
Densitet: 1,499
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00051697
CAS nummer: 2888-06-4
EINECS: 220-753-0
UN: 3265
ADR: 8,II