3,5-Dichlorobenzenesulphonyl chloride ≥97%
Leverantör: Thermo Scientific
Fara
Synonyms: 3,5-Dichlorobenzene-1-sulphonyl chloride
A11003.03 A11003.06 A11003.14
A11003.03EA 605 SEK
A11003.03 A11003.06 A11003.14
3,5-Dichlorobenzenesulphonyl chloride ≥97%
3,5-Dichlorobenzenesulphonyl chloride
Purity: 97% CAS No.: 705-21-5 Molecular Formula: C6H3Cl3O2S Formula Weight: 245.51 Flash Point: 110°(230°F) Melting Point: 30-33° Boiling Point: 158-160°/15mm Beilstein Reference: 1528657 UN No.: 3261 MDL No.: MFCD00051698
Formel: C₆H₃Cl₃O₂S
MW: 245,51 g/mol
Kokpunkt: 158…160 °C (15 mmHg)
Smältpunkt: 30…33 °C
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00051698
CAS nummer: 705-21-5
UN: 3261
ADR: 8,II