2,3-Dichlorobenzenesulphonyl chloride ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: 2,3-Dichlorobenzene-1-sulphonyl chloride
A12283.06 A12283.14
A12283.06EA 1625 SEK
A12283.06 A12283.14
2,3-Dichlorobenzenesulphonyl chloride ≥98%
2,3-Dichlorobenzenesulphonyl chloride
Purity: 98% CAS No.: 82417-45-6 Molecular Formula: C6H3Cl3O2S Formula Weight: 245.51 Melting Point: 57-62° Boiling Point: 131-133°/1mm UN No.: 3261 MDL No.: MFCD00051844
Formel: C₆H₃Cl₃O₂S
MW: 245,51 g/mol
Kokpunkt: 131…133 °C (1 mmHg)
Smältpunkt: 57…62 °C
Storage Temperature: Omgivande
MDL Number: MFCD00051844
CAS nummer: 82417-45-6
UN: 3261
ADR: 8,II