2-Chloro-4-nitrophenyl isothiocyanate ≥97%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 2-Chloro-1-isothiocyanato-4-nitrobenzene, 1-Chloro-2-isothiocyanato-5-nitrobenzene
L10898.03 L10898.06
L10898.03EA 455 SEK
L10898.03 L10898.06
2-Chloro-4-nitrophenyl isothiocyanate ≥97%
2-Chloro-4-nitrophenyl isothiocyanate
Purity: 97% CAS No.: 23165-64-2 Molecular Formula: C7H3ClN2O2S Formula Weight: 214.63 Odour: Irritating Melting Point: 95-99° Beilstein Reference: 2972987 UN No.: 2923 MDL No.: MFCD00041244
Formel: C₇H₃ClN₂O₂S
MW: 214.63 g/mol
Smältpunkt: 95…99 °C
Storage Temperature: Omgivande
MDL Number: MFCD00041244
CAS nummer: 23165-64-2
UN: 2923
ADR: 8 III