4-Chlorophenyl isothiocyanate ≥98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 4-Chlorophenylisothiocyanate, 1-Chloro-4-isothiocyanatobenzene
A12693.06 A12693.14 A12693.22
A12693.06EA 1150 SEK
A12693.06 A12693.14 A12693.22
4-Chlorophenyl isothiocyanate ≥98%
4-Chlorophenyl isothiocyanate
Purity: 98% CAS No.: 2131-55-7 Molecular Formula: C7H4ClNS Formula Weight: 169.64 Flash Point: 110°(230°F) Odour: Pungent Boiling Point: 135-136°/24mm Beilstein Reference: 471610 UN No.: 2923 MDL No.: MFCD00004810
Formel: C₇H₄ClNS
MW: 169.63 g/mol
Kokpunkt: 135…136 °C (24 mmHg)
Smältpunkt: 43…46 °C
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00004810
CAS nummer: 2131-55-7
EINECS: 218-358-3
UN: 2923
ADR: 8 III