2-Chlorophenyl isothiocyanate ≥97%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 1-Chloro-2-isothiocyanatobenzene
A18238.06 A18238.14
A18238.06EA 1525 SEK
A18238.06 A18238.14
2-Chlorophenyl isothiocyanate ≥97%
2-Chlorophenyl isothiocyanate
Purity: 97% CAS No.: 2740-81-0 Molecular Formula: C7H4ClNS Formula Weight: 169.64 Flash Point: 110°(230°F) Refractive Index: 1.6608 Odour: Pungent Boiling Point: 260-262° Beilstein Reference: 2081473 UN No.: 2922 MDL No.: MFCD00004801
Formel: C₇H₄ClNS
MW: 169.63 g/mol
Kokpunkt: 260…262 °C
Densitet: 1.287
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00004801
CAS nummer: 2740-81-0
UN: 2922
ADR: 8 III