3,4-Dichlorophenyl isothiocyanate

Leverantör: Thermo Scientific
Fara

Synonyms: 1,2-Dichloro-4-isothiocyanatobenzene, 3,4-Dichlorophenyl Isothiocyanate

SB01665EA
MAYBSB01665EAEA 0 SEK
MAYBSB01665EA
3,4-Dichlorophenyl isothiocyanate
3,4-Dichlorophenyl isothiocyanate
Formel: C₇H₃Cl₂NS
MW: 204,08 g/mol
Kokpunkt: 298 °C (1013 hPa)
Smältpunkt: 60…61 °C
Densitet: 1,422 g/cm³ (20 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00037828
CAS nummer: 6590-94-9
EINECS: 229-526-0
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR