2,4,6-Trichlorophenyl isocyanate

Leverantör: Thermo Scientific

Synonyms: Isocyanic acid 2,4,6-Trichlorophenyl ester

SB01903.1
MAYBSB01903.1EA 0 SEK
MAYBSB01903.1
2,4,6-Trichlorophenyl isocyanate
2,4,6-Trichlorophenyl isocyanate
Formel: C₇H₂Cl₃NO
MW: 222,46 g/mol
Kokpunkt: 101 °C (2 mmHg)
Smältpunkt: 66…67 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00013845
CAS nummer: 2505-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR