4-Klorofenylisocyanat ≥98%
Leverantör: Thermo Fisher Scientific
Fara
A14966.06 A14966.14 A14966.22
A14966.06EA 505 SEK
A14966.06 A14966.14 A14966.22
4-Klorofenylisocyanat ≥98%
4-Klorofenylisocyanat
Purity: 98% CAS No.: 104-12-1 Molecular Formula: C7H4ClNO Formula Weight: 153.57 Storage Temperature: 2°C to 8°C Flash Point: 90°(194°F) Refractive Index: 1.5618 Odour: Pungent Melting Point: 29-31° Boiling Point: 203-204° Beilstein Reference: 386235 UN No.: 2811 MDL No.: MFCD00002024
Formel: C₇H₄ClNO
MW: 153.57 g/mol
Kokpunkt: 203…204 °C
Smältpunkt: 29…31 °C
Densitet: 1.200
Flampunkt: 90 °C (194 °F)
Storage Temperature: Kylskåp
MDL Number: MFCD00002024
CAS nummer: 104-12-1
EINECS: 203-176-9
UN: 2811
ADR: 6.1 II