2-Chloro-4-fluorobenzenesulphonyl chloride

Leverantör: Thermo Scientific

Synonyms: 2-Chloro-4-fluorobenzene-1-sulphonyl chloride

TL00318DA TL00318EA
MAYBTL00318DAEA 0 SEK
MAYBTL00318DA MAYBTL00318EA
2-Chloro-4-fluorobenzenesulphonyl chloride
2-Chloro-4-fluorobenzenesulphonyl chloride
Formel: C₆H₃Cl₂FO₂S
MW: 229,06 g/mol
Kokpunkt: 88…90 °C (1013 hPa)
MDL Number: MFCD00052957
CAS nummer: 85958-57-2
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR