3-Chloro-4-fluorobenzamide

Leverantör: Thermo Scientific

BTB03706EA
MAYBBTB03706EAEA 0 SEK
MAYBBTB03706EA
3-Chloro-4-fluorobenzamide
3-Chloro-4-fluorobenzamide
Formel: C₇H₅ClFNO
MDL Number: MFCD00153092
CAS nummer: 701-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR