Ethyl-4-chlorobenzoate ≥98%
Leverantör: Thermo Scientific
Synonyms: Ethyl 4-chlorobenzoate, 4-Chlorobenzoic acid ethyl ester
A12540.18 A12540.30
A12540.18EA 1700 SEK
A12540.18 A12540.30
Ethyl-4-chlorobenzoate ≥98%
Ethyl-4-chlorobenzoate
Purity: 98+% CAS No.: 7335-27-5 Molecular Formula: C9H9ClO2 Formula Weight: 184.62 Refractive Index: 1.5240 Boiling Point: 237-239° MDL No.: MFCD00013645
Formel: C₉H₉ClO₂
MW: 184,62 g/mol
Kokpunkt: 237…239 °C
Densitet: 1,18
Storage Temperature: Omgivande
MDL Number: MFCD00013645
CAS nummer: 7335-27-5
EINECS: 230-844-7