Methyl-3-chlorobenzoate ≥98%
Leverantör: Thermo Fisher Scientific
B21371.14 B21371.22
B21371.14EA 1025 SEK
B21371.14 B21371.22
Methyl-3-chlorobenzoate ≥98%
Methyl-3-chlorobenzoate
Purity: 98% CAS No.: 2905-65-9 Molecular Formula: C8H7ClO2 Formula Weight: 170.60 Flash Point: 104°(219°F) Refractive Index: 1.5310 Melting Point: 20-22° Boiling Point: 99-101°/12mm Beilstein Reference: 2045126 Merck Reference: 14,2123 MDL No.: MFCD00000592
Formel: C₈H₇ClO₂
Kokpunkt: 99…101 °C (12 mmHg)
Smältpunkt: 20…22 °C
Densitet: 1,227
Flampunkt: 104 °C (219 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000592
CAS nummer: 2905-65-9
EINECS: 220-814-1
Merck Index: 14,2123