Methyl-2-chlorobenzoate ≥98%
Leverantör: Thermo Scientific
Synonyms: 2-Chlorobenzoic acid emthyl ester
A11237.14 A11237.22 A11237.36
A11237.14EA 895 SEK
A11237.14 A11237.22 A11237.36
Methyl-2-chlorobenzoate ≥98%
Methyl-2-chlorobenzoate
Purity: 98% CAS No.: 610-96-8 Molecular Formula: C8H7ClO2 Formula Weight: 170.60 Flash Point: 108°(226°F) Refractive Index: 1.5360 Boiling Point: 233-235° Beilstein Reference: 1364684 MDL No.: MFCD00016337
Formel: C₈H₇ClO₂
MW: 170,6 g/mol
Kokpunkt: 233…235 °C
Densitet: 1,191
Flampunkt: 108 °C (226 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00016337
CAS nummer: 610-96-8
EINECS: 210-242-0