2-Chlorobenzamide ≥98%

Leverantör: Thermo Fisher Scientific
Fara

A15706.14 A15706.22
A15706.14EA 885 SEK
A15706.14 A15706.22
2-Chlorobenzamide ≥98%
2-Chlorobenzamide

Purity: 98% CAS No.: 609-66-5 Molecular Formula: C7H6ClNO Formula Weight: 155.58 Melting Point: 140-144° Beilstein Reference: 508510

Formel: C₇H₆ClNO
Smältpunkt: 140…144 °C
Densitet: 1,34 g/cm³ (18 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00007973
CAS nummer: 609-66-5
EINECS: 210-195-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR